Learn More
XPNPEP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 10661-1-AP-150UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16827183
  • 385.92 CHF / 150 Mikroliter
Lager vorrätig
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.
Spezifikation
Spezifikation
XPNPEP1
Polyclonal
Unconjugated
XPNPEP1
Cytosolic aminopeptidase P, SAMP, Soluble aminopeptidase P, X Pro aminopeptidase 1, Xaa Pro aminopeptidase 1, XPNPEP, XPNPEP1, XPNPEPL, XPNPEPL1, XPP1
Rabbit
Antigen Affinity Chromatography
RUO
170750, 170751, 7511
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O54975, Q6P1B1, Q9NQW7
Xpnpep1
XPNPEP1 Fusion Protein Ag1015
150 μL
Primary
Human, Mouse, Rat