Learn More
TOM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25813-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene was identified as a target of the v-myb oncogene. The encoded protein shares its N-terminal domain in common with proteins associated with vesicular trafficking at the endosome. It is recruited to the endosomes by its interaction with endofin. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spezifikation
TOM1 | |
Polyclonal | |
Unconjugated | |
TOM1 | |
Target of Myb protein 1, TOM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10043, 21968 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60784, O88746 | |
TOM1 | |
TOM1 Fusion Protein Ag22931 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |