Learn More
RAB4A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10347-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The RAB4A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RAB4A. This antibody recognizes human, mouse, rat antigen. The RAB4A antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
RAB4A | |
Polyclonal | |
Unconjugated | |
20 μL | |
5867 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
200 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |