Learn More
LOC123688 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26070-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Catalyzes the GTP-dependent phosphorylation of 5- hydroxy-L-lysine.Spezifikation
LOC123688 | |
Polyclonal | |
Unconjugated | |
HYKK | |
5-hydroxy-L-lysine kinase, AGPHD1, Hydroxylysine kinase, HYKK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
123688, 235386 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2RU49, Q5U5V2 | |
Hykk | |
AGPHD1 Fusion Protein Ag23420 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |