Learn More
Density Standards (150 °C), Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C
81.00€
Specifications
Quantity | 60 mL |
---|---|
Certifications/Compliance | Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation |
Product Code | Brand | Density | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Density | Price | Quantity & Availability | |||||
13450539
|
Paragon Scientific
DEN150-01 |
0.7324 g/mL (150°C) |
81.00€
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13460539
|
Paragon Scientific
DEN150-02 |
0.7795 g/mL (150°C) |
81.00€
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13470539
|
Paragon Scientific
DEN150-03 |
0.8277 g/mL (150°C) |
81.00€
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols.
- Supplied in tamper evident packaging with a 2 year shelf life.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- Manufactured in the United Kingdom
Specifications
60 mL | |
Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- DEN150-01
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : Kerosine sweetened
Nominal values quoted