Learn More
pH 5.00 Buffer Solution at 25°C, Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
Description | Accuracy ± 0.01 pH units |
pH | 5.00 ±0.01 |
Physical Form | Liquid |
Quantity | 5 L |
Packaging | HDPE Bottle GL45 |