Learn More
NKX3-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 13069-1-AP-150UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16871104
  • 385.92 CHF / 150 Mikroliter
Lager vorrätig
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.
Spezifikation
Spezifikation
NKX3-1
Polyclonal
Unconjugated
NKX3-1
BAPX2, Homeobox protein Nkx 3.1, NK3 homeobox 1, NKX3, NKX3 1, NKX3.1, NKX3-1, NKX3A
Rabbit
Antigen Affinity Chromatography
RUO
18095, 305999, 4824
-20°C
Liquid
Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97436, Q99801
Nkx3-1
NKX3-1 Fusion Protein Ag4070
150 μL
Primary
Human, Mouse, Rat