Learn More
HSP60 Mouse anti-Human, CoraLite 594, Clone: 2F10E7, Proteintech
Mouse Monoclonal Antibody
Marke:  Proteintech CL594-66041-100UL
Artikelnummer. 16880936
  • 423.57 CHF / 100 Mikroliter
Lager vorrätig
Entdecken Sie weitere Sonderangebote
Beschreibung
The HSP60 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human HSP60. This antibody recognizes human, mouse, rat antigen. The HSP60 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spezifikation
Spezifikation
HSP60
Monoclonal
CoraLite 594
100 μL
3329
IgG1
Immunofluorescence
2F10E7
Mouse
Primary
Human, Rat, Mouse